Methanol ≥99% Ph. Eur., Macron Fine Chemicals™
Dodavatel: Avantor Performance Materials
|
Nebezpečí
|
Synonyms:
Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
7669-33EA
1630
CZK
7669-33
7669-38
7669-39
Methanol ≥99% Ph. Eur., Macron Fine Chemicals™
Methanol
Vzorec:
CH₃OH MW: 32.04 Bod varu: 64.6 Bod tání: -98 Bod vzplanutí: 11 Storage Temperature: Ambient |
MDL Number:
MFCD00004595 CAS číslo: 67-56-1 UN: 1230 ADR: 3 II Merck Index: 13,05984 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...