Tisk…
(+)-cis-Diltiazem hydrochloride ≥99% (by HPLC), Sigma-Aldrich®
Dodavatel: Merck
Varování
Synonyms: (2S,3S)-(+)-3-Acetoxy-2,3-dihydro-5-[2-(dimethylamino)ethyl]-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one Hydrochloride, Diltiazem Hydrochloride, (+)-cis-Diltiazem HCl, (+)-cis-3-Acetoxy-5-(2-dimethylaminoethyl)-2,3-dihydro-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one Hydrochloride, (2S,3S)-5-(2-(Dimethylamino)ethyl)-2-(4-methoxyphenyl)-4-oxo-2,3,4,5-tetrahydrobenzo[b][1,4]thiazepin-3-yl acetate hydrochloride
M-Clarity™ quality level = MQ200
D2521-10G D2521-1G D2521-5G
SIALD2521-10GEA 22090 CZK
SIALD2521-10G SIALD2521-1G SIALD2521-5G
(+)-cis-Diltiazem hydrochloride ≥99% (by HPLC), Sigma-Aldrich®
(+)-cis-Diltiazem hydrochloride
Blocks and slow down, L-type Ca2+ channels, regulates Ca2+ release from intracellular stores in neutrophils, stimulates 1,4-dihydropyridine binding to Ca2+ channels, also acts as coronary vasodilator.
Vzorec: C₂₂H₂₆N₂O₄S·HCl
MW: 450.98 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00069252
CAS číslo: 33286-22-5