Pimaricin ≥95%, crystalline solid
Dodavatel: Cayman Chemical
Synonyms:
Natamycin
Natamycin is a naturally-occurring macrolide polyene antifungal agent produced during fermentation by the bacterium S. natalensis, commonly found in soil. Used to treat fungal infections, including candida, aspergillus, cephalosporium, fusarium, and penicillium.
- Blocks fungal growth by binding specifically to ergosterol
- Used in the food industry as a natural preservative
Synonyms: Antibiotic A 5283, CL 12,625, delvocid, E 235, mycophyt, myprozine, pimaricin, synogil and tennecetin.
Vzorec:
C₃₃H₄₇NO₁₃ MW: 665.733 g/mol |
MDL Number:
MFCD00135085 CAS číslo: 7681-93-8 |
Výsledky specifikačního testu
Formulation | A crystalline solid |
λmax | 219, 290, 303, 317 nm |
Purity | ≥95% |
Storage temperature | −20 °C |
Stability | ≥2 years |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...