Tisk…
Graduated pipettes, BLAUBRAND®, type 1, Class AS, partial delivery, with DAkkS certificate
Dodavatel: Brand
BLAUBRAND®
BRNDDAKKS27731EA 2560 CZK
BRNDDAKKS27731 BRNDDAKKS27721 BRNDDAKKS27722 BRNDDAKKS27724 BRNDDAKKS27727 BRNDDAKKS27729
Graduated pipettes, BLAUBRAND®, type 1, Class AS, partial delivery, with DAkkS certificate
Pipety Pipety graduované
AR-Glas®.
  • Calibrated to deliver (TD, Ex); type 1: Zero on top
  • Marks and inscriptions in high contrast blue enamel
  • DAkks calibration certificate
  • Only pipettes marked (*) are suitable for cotton plugs
Length: 360 mm ±10 mm except for 25 ml pipettes: 450 mm ±10 mm.
Certifikace: DIN EN ISO 835. DE-M marking.