Copper chromite, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Copperchromium oxide
M-Clarity™ quality level = MQ200
209317-100G
209317-500G
SIAL209317-100GEA
2620
CZK
SIAL209317-100G
SIAL209317-500G
Copper chromite, Sigma-Aldrich®
Copper chromite
Used in battery manufacturing.
Synonym: Copper chromium oxide.
Vzorec:
Cr₂Cu₂O₅ MW: 311,079 g/mol |
MDL Number:
MFCD00044868 CAS číslo: 12053-18-8 |
Výsledky specifikačního testu
Form | Powder |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...