Tisk…

Poloxamer 124 (Pluronic® L44)

Dodavatel: Spectrum Chemical

Synonyms: Pluronic® L44, PEG-PPG-PEG

P1168-500MLG P1168-2.5LTGL P1168-200LTBL P1168-20LT
SPCMP1168-500MLGEA 6530 CZK
SPCMP1168-500MLG SPCMP1168-2.5LTGL SPCMP1168-200LTBL SPCMP1168-20LT
Poloxamer 124 (Pluronic® L44)
Poloxamer 124 (Pluronic® L44)

Poloxamer 124 has an amphiphilic structure (both hydrophilic and lipophilic) and is a useful surfactant for the formulation of cosmetics and pharmaceuticals because it increases miscibility.


Poloxamer 124 has also been used to control thickness (viscosity) of personal care products and dish soaps.

Vzorec: HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)a
MDL Number: MFCD00082049
CAS číslo: 9003-11-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR