Tisk…

Cerulenin 98, crystalline solid

Dodavatel: Cayman Chemical
Varování

10005647-10 10005647-5 10005647-50
CAYM10005647-10EA 8460 CZK
CAYM10005647-10 CAYM10005647-5 CAYM10005647-50
Cerulenin 98, crystalline solid
Cerulenin

A number of tumor cells and cell lines have been observed to have highly upregulated expression and activity of fatty acid synthase (FAS). Inhibition of FAS by the irreversible inhibitor cerulenin leads to cytotoxicity and apoptosis in human cancer cell lines, an effect believed to be mediated by the accumulation of malonyl-coenzyme A in cells with an upregulated FAS pathway. Cerulenin also leads to profound weight loss and feeding inhibition in both high-fat diet wild type obese and leptin-deficient ob/ob mice.

Vzorec: C₁₂H₁₇NO₃
MW: 223.272 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00077686
CAS číslo: 17397-89-6

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR