Tisk…

Cefsulodin sodium salt

Dodavatel: MP Biomedicals
Varování

Synonyms: Cefsulodin sodium, Sodium (6R,7R)-3-((4-carbamoylpyridin-1-ium-1-yl)methyl)-8-oxo-7-((R)-2-phenyl-2-sulphonatoacetamido)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate, Sulcephalosporin

0219867780 0219867783
ICNA0219867780EA 2740 CZK
ICNA0219867780 ICNA0219867783
Cefsulodin sodium salt
Cefsulodin sodium salt

Storage temperature.: +4 °C

Vzorec: C₂₂H₁₉N₄NaO₈S₂
MW: 554,524 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07793338
CAS číslo: 52152-93-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR