Tisk…

Thiamphenicol, crystalline

Dodavatel: MP Biomedicals

Synonyms: 2,2-Dichloro-N-[(1R,2R)-1,3-dihydroxy-1-[4-(methylsulphonyl)phenyl]-2-propyl]acetamide, (Methylsulphonyl)chloramphenicol

0215684605 0215684625
ICNA0215684605EA 5000 CZK
ICNA0215684605 ICNA0215684625
Thiamphenicol, crystalline
Thiamphenicol

Synonym: D-threo-2,2-dichloro-N-[β-hydroxy-α-(hydroxymethyl)-p-(methyl- sulfonyl)phenethyl]-acetamide

Vzorec: C₁₂H₁₅Cl₂NO₅S
MW: 356,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00467983
CAS číslo: 15318-45-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR