Tisk…
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)
Dodavatel: MP Biomedicals
Synonyms: Nadide phosphate disodium salt, Triphosphopyridine nucleotide, disodium salt, NADP-Na2, β-NADP-Na2, NADP disodium salt, Nicotinamide-adenine dinucleotide phosphate disodium salt, 1-((2R,3R,4S,5R)-5-((((((((2R,3R,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3-hydroxy-4-(phosphonooxy)tetrahydrofuran-2-yl)methoxy)(hydroxy)phosphoryl)oxy)oxidophosphoryl)oxy)methyl)-3,4-dihydroxytetrahydrofuran-2-yl)-3-carbamoylpyridin-1-ium, disodium salt, β-NADH phosphate disodium salt
0210116650 0210116680 0210116683 0210116691 0210116694
ICNA0210116650EA 2060 CZK
ICNA0210116650 ICNA0210116680 ICNA0210116683 ICNA0210116691 ICNA0210116694
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)
Nicotinamide adenine dinucleotide phosphate (NADP) and NADPH form a redox pair. NADP/NADPH is a coenzyme that supports redox reactions via the transport of electrons in a vast array of applications, especially anaerobic reactions such as lipid and nucleic acid synthesis. NADP/NADPH is a coenzyme couple in various cytochrome P450 systems and oxidase/reductase reaction systems, such as the thioredoxin reductase/thioredoxin system. This product will meet or exceed NRC specifications.
  • Coenzyme in aerobic and anaerobic oxidations
Storage temperature.: +4 °C, protect from light, under nitrogen.
White powder
NADP is an essential coenzyme for glucose-6-phosphate dehydrogenase which catalyses the oxidation of glucose-6-phosphate to 6-phosphogluconic acid. This reaction initiates metabolism of glucose by a pathway other than the citric acid cycle. This route is known as the hexose phosphate shunt or phosphogluconate pathway.
The nicotinamide portion of the coenzyme transfers hydrogens by alternating between oxidized quaternary nitrogen and a reduced tertiary nitrogen.
Vzorec: C₂₁H₂₆N₇Na₂O₁₇P₃
MW: 787.37
MDL Number: MFCD00065390
CAS číslo: 24292-60-2