Tisk…

Nitrilotris(methylenephosphonic acid) ≥97% (by titrimetric analysis), Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: Tris(phosphonomethyl)amine, Nitrilotri(methylphosphonic acid)

72568-10G 72568-50G
SIAL72568-10GEA 2800 CZK
SIAL72568-10G SIAL72568-50G
Nitrilotris(methylenephosphonic acid) ≥97% (by titrimetric analysis), Sigma-Aldrich®
Nitrilotris(methylenephosphonic acid)

Powerful complexing agent.

Vzorec: C₃H₁₂NO₉P₃
MW: 299,05 g/mol
Bod tání: ∼215 °C
MDL Number: MFCD00002138
CAS číslo: 6419-19-8

Objednat

Výsledky specifikačního testu

Appearance (Color) Colorless or White
Appearance (Form) Powder or Crystals
Assay by Titration (T) NaOH 1 M ≥97,0%
Proton NMR spectrum Conforms to Structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR