Tisk…
Centrifugační lahve, bio láhev, se šroubovacím uzávěrem, Nalgene®
Dodavatel: Thermo Fisher Scientific
3120-2006 3120-2024
525-2317EA 19410 CZK
525-2317 525-0987
Centrifugační lahve, bio láhev, se šroubovacím uzávěrem, Nalgene®
Láhve Lahvičky odstřeďovací
PPCO with white PP screw cap and silicone gasket.
  • Ergonomic design facilitates loading, recovery and storage of samples. Smooth inner walls are less likely to shear delicate cells
  • PPCO offers excellent chemical resistance
  • For this non sterile bottle maximum service life is 50 cycles
  • Recommended operating temperature range is from 4 to 22 °C; storage to 0 °C
  • Meets USP VI, is non cytotoxic and non haemolytic
Designed to process a 12 litre harvest in a single run when used in a Thermo Scientific Sorvall® RC12BP Centrifuge and H-12000 Swing Bucket Rotor. Replaces bio bottle. These bottles are ideal for bacterial, yeast and tissue isolations. Large volume gross separations up to 12 litres can be centrifuged up to 7333×g. Graduated, wide-mouth bottle simplifies decanting, re-suspensions and pellet removal.