Tisk…

(-)-1,2-Bis((2R,5R)-2,5-diphenylphospholano)ethane ≥97%

Dodavatel: Thermo Fisher Scientific

H60046.MD H60046.03
H60046.MDEA 6870 CZK
H60046.MD H60046.03
(-)-1,2-Bis((2R,5R)-2,5-diphenylphospholano)ethane ≥97%
(-)-1,2-Bis((2R,5R)-2,5-diphenylphospholano)ethane

Purity: 97+% CAS No.: 528565-79-9 Molecular Formula: C34H36P2 Formula Weight: 506.60 Melting Point: 143-147° MDL No.: MFCD07369027

Vzorec: C₃₄H₃₆P₂
Bod tání: 143…147 °C
Storage Temperature: Ambient
MDL Number: MFCD07369027
CAS číslo: 528565-79-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR