Tisk…

1,3-benzodioxole-5-sulphonyl chloride 95%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

H54755.MD H54755.03 H54755.06
H54755.MDEA 2160 CZK
H54755.MD H54755.03 H54755.06
1,3-benzodioxole-5-sulphonyl chloride 95%
1,3-benzodioxole-5-sulphonyl chloride

Purity: 95% CAS No.: 115010-10-1 Molecular Formula: C7H5ClO4S Formula Weight: 220.63 UN No.: 3261 MDL No.: MFCD04974524

Vzorec: C₇H₅ClO₄S
Storage Temperature: Ambient
MDL Number: MFCD04974524
CAS číslo: 115010-10-1
UN: 3261
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR