Tisk…
Chemical resistant overalls, MICROCHEM® 5000, model 111
Dodavatel: Ansell
OR50T-00111-03 OR50T-00111-04 OR50T-00111-05 OR50T-00111-06 OR50T-00111-07
ANSEOR50T-00111-03EA 25830 CZK
ANSEOR50T-00111-03 ANSEOR50T-00111-04 ANSEOR50T-00111-05 ANSEOR50T-00111-06 ANSEOR50T-00111-07
Chemical resistant overalls, MICROCHEM® 5000, model 111
Jednorázové oděvy
These overalls are made from a highly visible multi-layer fabric (external film barrier layer/non woven inner layer/core high barrier film layer) that is strong, durable and suitable for workers in extremely hazardous areas, including HAZMAT response teams.
  • Elasticated two-piece hood, outer cuffs, waist and ankles
  • Double cuff design to enable a spraytight connection with chemical protective gauntlets (additional taping or Glove Link is required)
  • Double zip system helps ensure a liquid tight seal without the need for additional taping
  • Ultrasonically welded and taped seams provide a high barrier to liquids and particulates
  • Barrier to numerous organic and inorganic chemicals and biological hazards
  • Bright orange colour for improved worker safety
  • Protection: >480 minutes breakthrough time against 14 of 15 chemicals listed in EN ISO 6529
Typical applications include: Chemical handling, oil and petrochemicals, pharmaceuticals, mining, agriculture, fire service, industrial and tank cleaning, HAZMAT emergency response, sewage purification installations.
Certifikace: Type 3-B/4-B/5-B, EN 14126, EN 1073-2, EN 1149-5; Cat. III
Suitable for Ex-Zones