Tisk…

1,2-Bis[(2S,5S)-2,5-dimethyl-1-phospholanyl]benzene ≥97%

Dodavatel: Thermo Scientific

Synonyms: (+)-1,2-Bis((2S,5S)-2,5-dimethylphospholano)benzene, (S,S)-Me-DUPHOS, 1,2-Bis((2S,5S)-2,5-dimethylphospholan-1-yl)benzene

H60647.MD H60647.03
H60647.MDEA 4570 CZK
H60647.MD H60647.03
1,2-Bis[(2S,5S)-2,5-dimethyl-1-phospholanyl]benzene ≥97%
1,2-Bis[(2S,5S)-2,5-dimethyl-1-phospholanyl]benzene

It is a DuPhos and BPE ligands which are highly efficient privileged ligands.

Vzorec: C₁₈H₂₈P₂
MW: 306,37 g/mol
Bod tání: 66…76 °C
Storage Temperature: Ambient
MDL Number: MFCD00142322
CAS číslo: 136735-95-0

Objednat

Výsledky specifikačního testu

Sensitivity Air Sensitive
Solubility Insoluble in water

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR