Tisk…
Metamizole ≥98%, crystalline solid
Dodavatel: Cayman Chemical
Synonyms: (1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)(methylamino)methanesulphonic acid sodium salt, Sulpyrine, Sodium (1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)(methylamino)methanesulphonate, Metamizole sodium salt, Metamizole (sodium salt), Dipyrone
15771-25 15771-50
CAYM15771-25EA 1970 CZK
CAYM15771-25 CAYM15771-50
Metamizole ≥98%, crystalline solid
Metamizole
Metamizole (sodium salt) is a nonopioid prodrug that is rapidly hydrolyzed to the metabolites 4-methylaminoantipyrine (MAA) and 4-aminoantipyrine (AA), which have analgesic and antipyretic effects. MAA has been shown to inhibit COX activity 8-9-fold more potently than AA (COX-1: IC50S= 2,6 and 20,8 µM, respectively, and COX-2: IC50S= 4,7 and 41,8 µM, respectively). These metabolites can be further acylated with arachidonic acid to form compounds that can bind to central and peripheral cannabinoid (CB1 and CB2) receptors and TRPV1 receptors at low micromolar concentrations.
Vzorec: C₁₃H₁₆N₃O₄S·Na
MW: 333.3 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00020783
CAS číslo: 68-89-3