Calf collagen (from skin), MP Biomedicals
Dodavatel: MP Biomedicals
0215002610
0215002680
0215002683
ICNA0215002610EA
1290
CZK
ICNA0215002610
ICNA0215002680
ICNA0215002683
Calf collagen (from skin), MP Biomedicals
Proteiny
Collagen is a group of naturally occurring proteins found in animals, especially in the flesh and connective tissues of vertebrates. It is a component of skin, bone, tendon, and other fibrous connective tissues.
- Soluble in 0,075 M Sodium citrate and in dilute acetic acid
- Suitable for gel formation, platelet aggregation, collagenase activity