Tisk…

Anti-CD38 Mouse Monoclonal Antibody (FITC) [clone: HIT2]

Dodavatel: Tonbo Biosciences
35-0389-T025 35-0389-T100
35-0389-T025EA 991 CZK
35-0389-T025 35-0389-T100
Anti-CD38 Mouse Monoclonal Antibody (FITC) [clone: HIT2]
Protilátky
The HIT2 antibody is specific for human CD38, a 45 kD type II transmembrane glycoprotein expressed on thymocytes, activated T and B cells, and monocytes as well as other non-hematopoietic cells. CD38 is an ectoenzyme that functions to catalyze the synthesis and hydrolysis of cyclic ADP-ribose and is involved in cell signaling, regulating cell adhesion, and activation. Additionally, CD38 has been shown to be a prognostic marker for some leukemias and other diseases.

Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: HIT2
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: Mouse IgG1, kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR