Tisk…
Swabs in tubes
Dodavatel: Copan
155C 160C 168C 159C 162C
710-0457EA 815 CZK
710-0457 710-0430 710-0461 710-0928 710-0929
Swabs in tubes
Tampóny odběrové Tampóny v tubách
Swabs in tubes manufactured from high quality PP.
  • Sterile
  • Each tube has a tamper-proof cap
  • Batch number, expiry date, and full description printed on each tube
Suitable for medical use guarantees that all swab tubes are non toxic and have low moisture permeability.
Upozornení: *Výrobky na této stránce nespadají do oblasti působnosti směrnice EU 2019/904