Tisk…
Hubbell plugout for industrial plug connections
Dodavatel: Brady
065968 065695
BRDY065968EA 2770 CZK
BRDY065968 BRDY065695
Hubbell plugout for industrial plug connections
Zámky bezpečnostní
These Hubbell plugouts effectively secure all larger industrial plug connections.
  • Two or four padlocks can be applied
Max. shackle diameter: 7,00 mm
Min. shackle clearance: 20,00 mm