Tisk…
Bottle top vacuum filtration systems, Corning®
Dodavatel: Corning
430186 430515 430516 430517 430756 430758 430767 430768 430769 430770 430771 430773 431096 431097 431098 431153 431154 431155 431205 431206 431474 431475
734-1821EA 8930 CZK
734-1821 734-1844 734-1845 734-1846 734-1860 734-1861 513-3346 513-3347 513-3348 513-3349 513-3350 513-3352 513-3354 513-3355 513-3356 513-3357 513-3358 513-3359 734-1900 734-1901 514-0168 514-0169
Bottle top vacuum filtration systems, Corning®
Filtry Nástavce filtrační
Vacuum filter systems available in four sizes: 150, 250, 500 ml and 1 L. Adapters are colour coded by membrane type for easy product identification: Polyethersulfone (PES) yellow; cellulose acetate (CA) orange; cellulose nitrate (CN) blue; nylon (NY) red.
  • Angled hose connector simplifies vacuum line attachment
  • Receiver bottles feature easy grip sides for improved handling
  • Individually packaged, sterile, certified non pyrogenic
  • Caps for receiver bottles are sterile and individually packaged
  • Extra plastic storage bottles are available
* 500 ml funnel with 70 mm membrane.
Informace pro objednávání: Systems are comprised of filter funnel, filter membrane, filter cap, neck adapter, receiver, receiver cap, and hose connector.