Tisk…

Anti-BrdU Mouse Monoclonal Antibody (Purified) [clone: MoBu-1]

Dodavatel: Biotium
BNUB0884-500 BNUB0884-100
BNUB0884-500EA 14540 CZK
BNUB0884-500 BNUB0884-100
Anti-BrdU Mouse Monoclonal Antibody (Purified) [clone: MoBu-1]
Protilátky
This antibody reacts with Bromodeoxyuridine (BrdU) in single stranded DNA (produced by partial denaturation of double stranded DNA), BrdU coupled to a protein carrier, as well as free BrdU. BrdU is a thymidine analog, incorporated into cell nuclei during DNA synthesis prior to mitosis. Antibody to BrdU is helpful in detecting S-phase cells, providing useful information on the aggressiveness of tumors.

Type: Primary
Antigen: Not Applicable
Clonality: Monoclonal
Clone: MoBu-1
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: All species
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR