Tisk…

Anti-CDKN1B Mouse Monoclonal Antibody (CF405S) [clone: SX53G8]

Dodavatel: Biotium
BNC041237-500 BNC041237-100
BNC041237-500EA 15940 CZK
BNC041237-500 BNC041237-100
Anti-CDKN1B Mouse Monoclonal Antibody (CF405S) [clone: SX53G8]
Protilátky
This antibody recognizes a 27 kDa protein, identified as the p27Kip1, a cell cycle regulatory mitotic inhibitor. Its epitope spans between aa 83-204 of p27. It is highly specific and shows no cross-reaction with other related mitotic inhibitors. p27Kip1 functions as a negative regulator of G1 progression and has been proposed to function as a possible mediator of TGF- induced G1 arrest. p27Kip1 is a candidate tumor suppressor gene. This MAb co-precipitates cdk4 in complex p27Kip1 and is excellent for staining of formalin-fixed tissues.

Type: Primary
Antigen: CDKN1B
Clonality: Monoclonal
More Product Information
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR