Tisk…

Anti-PTPRC Mouse Monoclonal Antibody (CF488A) [clone: UCHL-1 T200/797]

Dodavatel: Biotium
BNC881209-500 BNC881209-100
BNC881209-500EA 15090 CZK
BNC881209-500 BNC881209-100
Anti-PTPRC Mouse Monoclonal Antibody (CF488A) [clone: UCHL-1 T200/797]
Protilátky
This antibody recognizes a 180-185 kDa protein, identified as isoform of leukocyte common antigen (CD45RO). This antibody reacts with mature activated T-cells, most thymocytes, and a sub-population of resting T-cells within both CD4 and CD8 subsets. It shows no reactivity with normal B or natural killer cells, but reacts with granulocytes and monocytes. Reportedly, it is useful to identify T-cell lymphomas and leukemias. It rarely stains NK cells or B-cell lymphomas.

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: UCHL-1 T200/797
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR