Tisk…

Anti-KRT7 Mouse Monoclonal Antibody (Biotin) [clone: KRT7/1198]

Dodavatel: Biotium
BNCB1198-500 BNCB1198-100
BNCB1198-500EA 15940 CZK
BNCB1198-500 BNCB1198-100
Anti-KRT7 Mouse Monoclonal Antibody (Biotin) [clone: KRT7/1198]
Protilátky
This antibody recognizes an intermediate filament protein (IFP) of 55 kDa, which is identified as cytokeratin 7. This MAb is highly specific to cytokeratin 7 and shows no cross-reaction with other IFPs. Cytokeratin 7 is a basic cytokeratin, which is found in most glandular and transitional epithelia but not in the stratified squamous epithelia. Keratin 7 is expressed in the epithelial cells of ovary, lung, and breast but not of colon, prostate, or gastrointestinal tract. This MAb is highly useful in distinguishing ovarian carcinomas (keratin 7 ) from colon carcinomas (keratin 7-).

Type: Primary
Antigen: KRT7
Clonality: Monoclonal
Clone: KRT7/1198
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR