Tisk…
Anti-CD66 molecule Mouse Monoclonal Antibody (CF647) [clone: C66/195 C66/261 C66/1009 C66/1030]
Dodavatel: Biotium
BNC471139-500 BNC471139-100
BNC471139-500EA 15940 CZK
BNC471139-500 BNC471139-100
Anti-CD66 molecule Mouse Monoclonal Antibody (CF647) [clone: C66/195 C66/261 C66/1009 C66/1030]
Protilátky
This antibody recognizes proteins of 80-200 kDa, identified as different members of CEA family. CEA is synthesized during development in the fetal gut and is re-expressed in increased amounts in intestinal carcinomas and several other tumors. This MAb does not react with nonspecific cross-reacting antigen (NCA) and with human polymorphonuclear leucocytes. It shows no reaction with a variety of normal tissues and is suitable for staining of formalin/paraffin tissues. CEA is not found in benign glands, stroma, or malignant prostatic cells. Antibody to CEA is useful in detecting early foci ofMore Product Information
Objednat
Produktová specifikace
Symbol antigenu CD66 molecule
Název antigenu CD66
Environmentally Preferable
Synonyma antigenu CEACAM5|CD66|Biliary Glycoprotein (BGP-1)|Carcinoembryonic Antigen-related Cell Adhesion Molecule 5
Druh protilátky Primární
Klonovalita Monoclonal
Spojení CF® 647
Klon Clone C66/195 C66/261 C66/1009 C66/1030
Reaktivita Human
Hostitel Mouse
Identifikační číslo genu 709196
Izotyp IgG
Imunochemie Yes
ImunoFluorescence Yes
Koncentrace 0,1 mg/ml
Skladovací pufr PBS/0,1% BSA/0,05% azide