Tisk…
Anti-Ep-CAM Mouse Monoclonal Antibody (CF647) [clone: EGP40/1120]
Dodavatel: Biotium
BNC471120-500 BNC471120-100
BNC471120-500EA 15940 CZK
BNC471120-500 BNC471120-100
Anti-Ep-CAM Mouse Monoclonal Antibody (CF647) [clone: EGP40/1120]
Protilátky
EGP40 is a 40-43 kDa transmembrane epithelial glycoprotein, also identified as epithelial specific antigen (ESA), or epithelial cellular adhesion molecule (Ep-CAM). It is expressed on baso-lateral cell surface in most simple epithelia and a vast majority of carcinomas. This antibody has been used to distinguish adenocarcinoma from pleural mesothelioma and hepatocellular carcinoma. This antibody is also useful in distinguishing serous carcinomas of the ovary from mesothelioma.
Type: Primary
Antigen: Ep-CAM
Clonality: Monoclonal
Clone: EGP40/1120
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human