Tisk…
Anti-HLA-A Mouse Monoclonal Antibody (CF647) [clone: 108-2C5]
Dodavatel: Biotium
BNC471095-500 BNC471095-100
BNC471095-500EA 15940 CZK
BNC471095-500 BNC471095-100
Anti-HLA-A Mouse Monoclonal Antibody (CF647) [clone: 108-2C5]
Protilátky
HLA-A, with HLA-B and HLA-C, belongs to major histocompatibility complex (MHC) class I antigens and expresses constitutively on all nucleated cells. MHC class I antigens play a role in class I MHC-associated antigen presentation, inhibition of NK cell cytotoxicity, tumor surveillance, and tissue allotransplantation. This MAb is useful for HLA molecular typing of peripheral blood leukocytes as well as a large number of leukemic cell lines. It reacts with an intralocus determinant present on a limited number of HLA-A locus-encoded gene products (HLA-A2, -A3, -A28, -A29, -A30, -A31 and -Aw33).More Product Information