Tisk…
Anti-HLA-A Mouse Monoclonal Antibody (CF568) [clone: CATA-1]
Dodavatel: Biotium
BNC681073-500 BNC681073-100
BNC681073-500EA 15940 CZK
BNC681073-500 BNC681073-100
Anti-HLA-A Mouse Monoclonal Antibody (CF568) [clone: CATA-1]
Protilátky
This MAb reacts with cells bearing HLA-A25 or HLA-Aw32 antigens. In addition, a reaction was observed with a cell of phenotype A2, Aw31; B17, Bw49. HLA-A, with HLA-B and HLA-C, belongs to major histocompatibility complex (MHC) class I antigens and expresses constitutively on all nucleated cells. HLA system comprises closely linked genes controlling highly polymorphic proteins involved in the presentation of peptides to the T-cell receptor, inhibition of NK cell cytotoxicity, and rejection of tissue allotransplantation. Specific alleles at HLA loci are associated with diseases. This MAb isMore Product Information