Tisk…

Anti-VEGFA Mouse Monoclonal Antibody (CF488A) [clone: VEGF/1063]

Dodavatel: Biotium
BNC881063-500 BNC881063-100
BNC881063-500EA 15940 CZK
BNC881063-500 BNC881063-100
Anti-VEGFA Mouse Monoclonal Antibody (CF488A) [clone: VEGF/1063]
Protilátky
This MAb recognizes proteins of 19-22 kDa (reducing) and 38 kDa-44 kDa (non-reducing), identified as various isoforms of Vascular Endothelial Growth Factor or Vascular Permeability Factor (VEGF/VPF). It is highly specific to VEGF, which is a homodimeric, disulfide-linked glycoprotein with a close homology to platelet derived growth factor (PDGF). There are multiple isoforms of VEGF containing 206-, 189-, 165-, and 121-amino acid residues. The smaller two isoforms, VEGF165 and VEGF121, are secreted proteins and act as diffusible agents, whereas the larger two remain cell associated. VEGF/VPF plays an important role in angiogenesis, which promotes tumor progression and metastasis.

Type: Primary
Antigen: VEGFA
Clonality: Monoclonal
Clone: VEGF/1063
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Dog, Human, Mouse, Rabbit, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR