Tisk…

Anti-HLA-A; HLA-B; HLA-C Mouse Monoclonal Antibody (CF488A) [clone: 246-B8.E7]

Dodavatel: Biotium
BNC881054-500 BNC881054-100
BNC881054-500EA 15940 CZK
BNC881054-500 BNC881054-100
Anti-HLA-A; HLA-B; HLA-C Mouse Monoclonal Antibody (CF488A) [clone: 246-B8.E7]
Reacts with a monomorphic determinant of human major histocompatibility (MHC) class I antigens (HLA-A, B and C). Human MHC class I antigens are expressed constitutively on all nucleated cells lymphocytes such as lymphocytes, thymocytes, granulocytes, and bone marrow cells and are absent on erythrocytes. MHC class I antigens play a role in class I MHC-associated antigen presentation, inhibition of NK cell cytotoxicity, tumor surveillance, and tissue allotransplantation.

Type: Primary
Antigen: HLA-A; HLA-B; HLA-C
Clonality: Monoclonal
Clone: 246-B8.E7
Conjugation: CF488A
Epitope:
Host: Mouse
More Product Information
Objednat

Produktová specifikace

Symbol antigenu MHC I
Environmentally Preferable
Název antigenu MHC Class 1
Synonyma antigenu HLA-B|HLA-A|HLA-C
Druh protilátky Primární
Klonovalita Monoclonal
Spojení CF® 488A
Klon Clone 246-B8.E7
Reaktivita Primates,Human,Mouse
Hostitel Mouse
Identifikační číslo genu 181244; 654404; 77961
Izotyp IgG2a kappa
ImunoFluorescence Yes
Koncentrace 0,1 mg/ml
Skladovací pufr PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR