Tisk…
Anti-S100A1 Mouse Monoclonal Antibody (CF405S) [clone: S100A1/1012]
Dodavatel: Biotium
BNC041012-500 BNC041012-100
BNC041012-500EA 15940 CZK
BNC041012-500 BNC041012-100
Anti-S100A1 Mouse Monoclonal Antibody (CF405S) [clone: S100A1/1012]
Protilátky
The family of EF-hand type Ca2 -binding proteins includes calbindin, S-100 alpha and beta, calgranulins, B and C, and the parvalbumin family members, including parvalbumin alpha and parvalbumin beta. The S-100 protein is involved in the regulation of cellular processes such as cell cycle progression and differentiation. S-100 protein may function in the activation of Ca2 induced Ca2 release, inhibition of microtubule assembly and inhibition of protein kinase C mediated phosphorylation. Two S-100 subunits, sharing 60% sequence identity, have been described as S-100 alpha chain and S-100 betaMore Product Information
Objednat
Produktová specifikace
Symbol antigenu S100A1
Název antigenu S100
Environmentally Preferable
Synonyma antigenu S100 Calcium Binding Protein A1|Bpb|S100 Alpha Chain|S100 Calcium Binding Protein B|NEF|S100-A1|S100 Beta Chain|S100 Calcium Binding Protein Beta Neural
Druh protilátky Primární
Klonovalita Monoclonal
Spojení CF® 405S
Klon Clone S100A1/1012
Reaktivita Human,Bovine,Rat,Mouse
Hostitel Mouse
Identifikační číslo genu 515715
Izotyp IgG2a kappa
Western blot Yes
Imunochemie Yes
ImunoFluorescence Yes
Koncentrace 0,1 mg/ml
Skladovací pufr PBS/0,1% BSA/0,05% azide