Tisk…

Anti-P62 Rabbit Polyclonal Antibody

Dodavatel: Novus Biologicals
NBP1-48320 NBP1-48320SS
NOVUNBP1-48320EA 19680 CZK
NOVUNBP1-48320 NOVUNBP1-48320SS
Anti-P62 Rabbit Polyclonal Antibody
Protilátky
Rabbit Polyclonal p62/SQSTM1 Antibody. Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence. Tested Reactivity: Human, Mouse, Rat, Sheep.

There is 1 scientific publication using this antibody.

Type: Primary
Antigen: P62
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR