Tisk…

Anti-TNF Mouse Monoclonal Antibody (CF647) [clone: J1D9]

Dodavatel: Biotium
BNC470993-500 BNC470993-100
BNC470993-500EA 15940 CZK
BNC470993-500 BNC470993-100
Anti-TNF Mouse Monoclonal Antibody (CF647) [clone: J1D9]
Protilátky
This antibody neutralizes TNF alpha biological activities. It prevents TNF alpha induced apoptosis in Jurkat cells. It also neutralizes HurTNFamediated cytotoxicity of L929 cells and inhibits tumor growth in mice. It protects mice against toxicity of HurTNFa. Tumor Necrosis Factor Alpha (TNF alpha) is a protein secreted by lipopolysaccharide-stimulated macrophages, and causes tumor necrosis when injected into tumor bearing mice. TNF alpha is believed to mediate pathogenic shock and tissue injury associated with endotoxemia. TNF alpha exists as a multimer of two, three, or five non-covalently linked units, but shows a single 17 kDa band following SDS PAGE under non-reducing conditions. TNF alpha is closely related to the 25 kDa protein Tumor Necrosis Factor beta (lymphotoxin), sharing the same receptors and cellular actions. TNF alpha causes cytolysis of certain transformed cells, being synergistic with interferon gamma in its cytotoxicity. Although it has little effect on many cultured normal human cells, TNF alpha appears to be directly toxic to vascular endothelial cells. Other actions of TNF alpha include stimulating growth of human fibroblasts and other cell lines, activating polymorphonuclear neutrophils and osteoclasts, and induction of interleukin 1, prostaglandin E2 and collagenase production.

Type: Primary
Antigen: TNF
Clonality: Monoclonal
Clone: J1D9
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR