Tisk…
Anti-MUC3A Mouse Monoclonal Antibody (CF405S) [clone: M3.1]
Dodavatel: Biotium
BNC040990-500 BNC040990-100
BNC040990-500EA 15940 CZK
BNC040990-500 BNC040990-100
Anti-MUC3A Mouse Monoclonal Antibody (CF405S) [clone: M3.1]
Protilátky
This antibody recognizes a protein of HMW, identified as mucin 3 glycoprotein (MUC3). Its epitope localizes between aa SITTTE. This MAb shows no cross-reaction with human milk fat globule membranes, MUC1, or MUC2. MUC3 is distributed in colon and rectum, and is also present to a lesser extent in breast, lung and salivary gland tissues. The Mucins are a family of highly glycosylated, secreted proteins with a basic structure consisting of a variable number of tandem repeats (VNTRs) encoded by 60 base pairs (Mucin 1), 69 base pairs (Mucin 2) and 51 base pairs (Mucin 3). The number of repeats isMore Product Information