Tisk…
Anti-CD7 Mouse Monoclonal Antibody (CF647) [clone: T3-3A1]
Dodavatel: Biotium
BNC470978-500 BNC470978-100
BNC470978-500EA 15940 CZK
BNC470978-500 BNC470978-100
Anti-CD7 Mouse Monoclonal Antibody (CF647) [clone: T3-3A1]
Protilátky
Recognizes a protein of 40 kDa, identified as CD7, a member of the immunoglobulin gene superfamily. Its N-terminal amino acids 1-107 are highly homologous to Ig kappa-L chains whereas the carboxyl-terminal region of the extracellular domain is proline-rich and has been postulated to form a stalk from which the Ig domain projects. CD7 is expressed on the majority of immature and mature T-lymphocytes, and T cell leukemia. It is also found on natural killer cells, a small subpopulation of normal B cells and on malignant B cells. Cross-linking surface CD7 positively modulates T cell and NK cellMore Product Information