Tisk…
Anti-TFGB Mouse Monoclonal Antibody (CF488A) [clone: 1D11.16.8]
Dodavatel: Biotium
BNC880977-500 BNC880977-100
BNC880977-500EA 15940 CZK
BNC880977-500 BNC880977-100
Anti-TFGB Mouse Monoclonal Antibody (CF488A) [clone: 1D11.16.8]
Protilátky
This MAb recognizes TGF beta 1, 2 and 3. Three TGF betas have been identified in mammals. TGF beta-1, TGF beta-2 and TGF beta-3 are each synthesized as precursor proteins that are very similar in that each is cleaved to yield a 112 amino acid polypeptide that remains associated with the latent portion of the molecules. Biologically active TGF betarequires dimerization of the monomers (usually homodimers) and release of the latent peptide portion. Overall, the mature region of the TGF beta-3 protein has approximately 80% identity to the mature region of both TGF beta-1 and TGF beta-2. However,More Product Information