Tisk…

Anti-MYADM Mouse Monoclonal Antibody (CF640R) [clone: MYADM/971]

Dodavatel: Biotium
BNC400971-500 BNC400971-100
BNC400971-500EA 15940 CZK
BNC400971-500 BNC400971-100
Anti-MYADM Mouse Monoclonal Antibody (CF640R) [clone: MYADM/971]
Protilátky
Recognizes a myeloid associated differentiation antigen in the cytoplasm of mature granulocytes. It shows no reactivity with any other cell type in human tissues. Markers of myeloid cells are useful in the identification of different levels of cellular differentiation. It reacts with early precursor and mature forms of human and monkey myeloid cells. This MAb is useful for the detection of myeloid leukemias and granulocytic sarcomas. It can be used as a marker of granulocytes in normal tissues or inflammatory processes.

Type: Primary
Antigen: MYADM
Clonality: Monoclonal
Clone: MYADM/971
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Macaque
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR