Tisk…

Anti-MFGE8 Mouse Monoclonal Antibody (CF647) [clone: EDM45]

Dodavatel: Biotium
BNC470942-500 BNC470942-100
BNC470942-500EA 15940 CZK
BNC470942-500 BNC470942-100
Anti-MFGE8 Mouse Monoclonal Antibody (CF647) [clone: EDM45]
Protilátky
Recognizes a protein of 40-45 kDa, identified as human milk fat globule membrane protein (HMFG). HMFG is present on normal human breast epithelial cells and cell lines derived from breast carcinomas, as well as to the outer surface of the human milk fat globule. HMFG is considered as a differentiation marker. It is useful as specific breast epithelial marker and can also provide a tool to study the role of the cell surface in normal and neoplastic mammary development.

Type: Primary
Antigen: MFGE8
Clonality: Monoclonal
Clone: EDM45
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR