Tisk…

Anti-MFGE8 Mouse Monoclonal Antibody (Purified) [clone: EDM45]

Dodavatel: Biotium
BNUB0942-500 BNUB0942-100
BNUB0942-500EA 14540 CZK
BNUB0942-500 BNUB0942-100
Anti-MFGE8 Mouse Monoclonal Antibody (Purified) [clone: EDM45]
Protilátky
Recognizes a protein of 40-45 kDa, identified as human milk fat globule membrane protein (HMFG). HMFG is present on normal human breast epithelial cells and cell lines derived from breast carcinomas, as well as to the outer surface of the human milk fat globule. HMFG is considered as a differentiation marker. It is useful as specific breast epithelial marker and can also provide a tool to study the role of the cell surface in normal and neoplastic mammary development.

Type: Primary
Antigen: MFGE8
Clonality: Monoclonal
Clone: EDM45
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR