Tisk…

Anti-MUC6 Mouse Monoclonal Antibody (CF647) [clone: MUC6/916]

Dodavatel: Biotium
BNC470916-500 BNC470916-100
BNC470916-500EA 15940 CZK
BNC470916-500 BNC470916-100
Anti-MUC6 Mouse Monoclonal Antibody (CF647) [clone: MUC6/916]
Protilátky
The MUC6 gastric mucin is a secreted glycoprotein that plays an essential role in epithelial cyto-protection from acid, proteases, pathogenic microorganisms, and mechanical trauma in the gastrointestinal tract. Mucin 6 expression is highest in the stomach and gall bladder, with lower expression in the terminal ileum and right colon. In gastric cancer, Mucin 6 has an altered expression. In normal stomach, Mucin 6 is associated with Lewis type 2; Mucin 6 is also expressed in gastric metaplasia, duodenum and pancreas. Mucin 6 is a secretory mucin, located in the deeper mucosal folds of human gall bladder, and its expression is altered with increasing degrees of inflammation.

Type: Primary
Antigen: MUC6
Clonality: Monoclonal
Clone: MUC6/916
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR