Tisk…

Anti-KRT7 Mouse Monoclonal Antibody (CF594) [clone: KRT7/903]

Dodavatel: Biotium
BNC940903-100 BNC940903-500
BNC940903-100EA 5770 CZK
BNC940903-100 BNC940903-500
Anti-KRT7 Mouse Monoclonal Antibody (CF594) [clone: KRT7/903]
Protilátky
Cytokeratin 7 is a basic cytokeratin, which is found in most glandular and transitional epithelia but not in the stratified squamous epithelia. Keratin 7 is expressed in the epithelial cells of ovary, lung, and breast but not of colon, prostate, or gastrointestinal tract. Antibody to cytokeratin is useful in distinguishing ovarian carcinomas (keratin 7 ) from colon carcinomas (keratin 7-).

Type: Primary
Antigen: KRT7
Clonality: Monoclonal
Clone: KRT7/903
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR