Tisk…
Anti-PNL2 Mouse Monoclonal Antibody (CF660R) [clone: PNL2]
Dodavatel: Biotium
BNC610894-500 BNC610894-100
BNC610894-500EA 15940 CZK
BNC610894-500 BNC610894-100
Anti-PNL2 Mouse Monoclonal Antibody (CF660R) [clone: PNL2]
Protilátky
Anti-PNL2 is a novel monoclonal antibody, which has recently been introduced as an immunohistochemical reagent to stain melanocytes and tumors derived therefrom. The antigen recognized by PNL2 is different from Melan A and gp100. Its epitope is not destroyed by digestion with neuraminidase i.e. its epitope id not glycosylated. Anti-PNL2 may be most useful because of its high sensitivity for metastatic melanoma (87%), as opposed to 76% for anti-HMB45 and 82% for anti-MART-1. Anti-PNL2 labels intra-epidermal nevi while the dermal component of compound nevi are largely non-reactive with anti-More Product Information