Tisk…
Anti-AR Mouse Monoclonal Antibody (Biotin) [clone: AR441]
Dodavatel: Biotium
BNCB0881-500 BNCB0881-100
BNCB0881-500EA 15940 CZK
BNCB0881-500 BNCB0881-100
Anti-AR Mouse Monoclonal Antibody (Biotin) [clone: AR441]
Protilátky
Recognizes a protein of 110 kDa, which is identified as androgen receptor (AR). It reacts with full length, and the newly described A form of the receptor. It does not cross react with estrogen, progesterone, or glucocorticoid receptors. The expression of AR is reportedly inversely correlated with histologic grade i.e. well differentiated prostate tumors show higher expression than the poorly differentiated tumors. In prostate cancer, AR has been proposed, as a marker of hormone-responsiveness and thus it may be useful in identifying patients likely to benefit from anti-androgen therapy.More Product Information