Tisk…
Anti-CDKN1C Mouse Monoclonal Antibody (CF488A) [clone: KP10]
Dodavatel: Biotium
BNC880879-500 BNC880879-100
BNC880879-500EA 15940 CZK
BNC880879-500 BNC880879-100
Anti-CDKN1C Mouse Monoclonal Antibody (CF488A) [clone: KP10]
Protilátky
Recognizes a protein of 57 kDa, identified as p57Kip2. It shows no cross-reaction with p27Kip1. p57Kip2 is a potent tight-binding inhibitor of several G1 cyclin complexes, and is a negative regulator of cell proliferation. Anti-p57 has been used as an aide in identification of complete hydatidiform mole (CHM) (no nuclear labeling of cytotrophoblasts and stromal cells) from partial hydatidiform mole (PHM) in which both cytotrophoblasts and stromal cells stain. The histological differentiation of complete mole, partial mole, and hydropic spontaneous abortion is problematic. Most completeMore Product Information
Objednat
Produktová specifikace
Symbol antigenu CDKN1C
Environmentally Preferable
Název antigenu Cyclin-dependent kinase inhibitor 1C
Synonyma antigenu BWCR|p57|Cyclin dependent kinase inhibitor 1C (CDKN1C)|Beckwith Wiedemann syndrome (WBS)|p57Kip2|KIP2|Cyclin dependent kinase inhibitor p57
Druh protilátky Primární
Klonovalita Monoclonal
Spojení CF® 488A
Klon Clone KP10
Reaktivita Human,Mouse
Hostitel Mouse
Identifikační číslo genu 106070
Izotyp IgG2b kappa
Imunochemie Yes
ImunoFluorescence Yes
Koncentrace 0,1 mg/ml
Skladovací pufr PBS/0,1% BSA/0,05% azide