Tisk…
Anti-HSPD1 Mouse Monoclonal Antibody (CF488A) [clone: HSPD1/875]
Dodavatel: Biotium
BNC880875-500 BNC880875-100
BNC880875-500EA 15940 CZK
BNC880875-500 BNC880875-100
Anti-HSPD1 Mouse Monoclonal Antibody (CF488A) [clone: HSPD1/875]
Protilátky
Recognizes a 60 kDa protein, identified as the heat shock protein 60 (hsp60). A wide variety of environmental and pathophysiological stressful conditions trigger the synthesis of a family of proteins known as heat shock proteins (hsp), more appropriately called as stress response proteins (srp). hsp60 is a potential antigen in a number of autoimmune diseases. In human arthritis and in experimentally induced arthritis in animals, disease development coincides with the development of immune reactivity directed against not only bacterial hsp60, but also against its mammalian homolog.