Tisk…

Anti-MUC2 Mouse Monoclonal Antibody (Purified) [clone: MLP/842]

Dodavatel: Biotium
BNUB0842-500 BNUB0842-100
BNUB0842-500EA 14540 CZK
BNUB0842-500 BNUB0842-100
Anti-MUC2 Mouse Monoclonal Antibody (Purified) [clone: MLP/842]
Protilátky
Recognizes a single glycoprotein of 520 kDa, identified as mucin 2 (MUC2). This MAb shows no cross-reaction with human milk fat globule membranes, MUC1, or MUC3. Mucins are high molecular weight glycoproteins, which constitute the major component of the mucus layer that protects the gastric epithelium.MUC2 is specifically expressed in goblet cells of the small intestine & colon; in about 65% of colonic carcinomas and about 40% of gastric carcinomas. MUC2 is rarely expressed outside of the GI tract with the exceptions of mucinous carcinoma of breast and clear cell-type carcinomas of the ovary.

Type: Primary
Antigen: MUC2
Clonality: Monoclonal
Clone: MLP/842
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR