Tisk…
Anti-MAGEA1 Mouse Monoclonal Antibody (CF488A) [clone: MZ2E/838]
Dodavatel: Biotium
BNC880838-500 BNC880838-100
BNC880838-500EA 15940 CZK
BNC880838-500 BNC880838-100
Anti-MAGEA1 Mouse Monoclonal Antibody (CF488A) [clone: MZ2E/838]
Protilátky
Recognizes a protein of 42-46 kDa, identified as MAGE-1. This MAb does not cross-react with other members of MAGE-family. Human malignant neoplasms carry rejection antigens that are recognized by the patients' autologous, tumor directed and specific, cytolytic, CD8 T lymphocyte clones (CTL). The MAGE family of genes codes an important group of antigens. It was identified that melanomas and primary glial brain tumors express common melanoma associated antigens (MAAs). Because MAGE-1 is expressed on a significant proportion of human neoplasms of various histological types (melanoma, brainMore Product Information