Tisk…

Anti-Ep-CAM Mouse Monoclonal Antibody (CF647) [clone: EGP40/826]

Dodavatel: Biotium
BNC470826-500 BNC470826-100
BNC470826-500EA 15940 CZK
BNC470826-500 BNC470826-100
Anti-Ep-CAM Mouse Monoclonal Antibody (CF647) [clone: EGP40/826]
Protilátky
Recognizes a 40-43 kDa transmembrane epithelial glycoprotein, identified as epithelial specific antigen (ESA), or epithelial cellular adhesion molecule (Ep-CAM). Ep-CAM is expressed on baso-lateral cell surface in most simple epithelia and a vast majority of carcinomas. This antibody has been used to distinguish adenocarcinoma from pleural mesothelioma and hepatocellular carcinoma. It is also useful in distinguishing serous carcinomas of the ovary from mesothelioma. This epithelial antigen plays an important role as a tumor-cell marker in lymph nodes from patients with esophageal carcinoma otherwise classified as node-negative. Epithelial antigen has also been suggested as a discriminator between basal cell and baso-squamous carcinomas, and squamous cell carcinoma of the skin.

Type: Primary
Antigen: Ep-CAM
Clonality: Monoclonal
Clone: EGP40/826
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR